Skip to Content
Merck
All Photos(3)

Key Documents

46093

Sigma-Aldrich

1-Ethyl-3-methylimidazolium hexafluorophosphate

≥97.0% (HPLC)

Synonym(s):

EMIMPF6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11F6N2P
CAS Number:
Molecular Weight:
256.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥97.0% (HPLC)

form

solid

mp

58-62 °C (lit.)
58-62 °C

SMILES string

F[P-](F)(F)(F)(F)F.CCn1cc[n+](C)c1

InChI

1S/C6H11N2.F6P/c1-3-8-5-4-7(2)6-8;1-7(2,3,4,5)6/h4-6H,3H2,1-2H3;/q+1;-1

InChI key

DPDAKOVGQUGTHH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Ethyl-3-methylimidazolium hexafluorophosphate is a task-specific ionic liquid (TSIL)[1] with low melting point.[2]

Other Notes

Air- and water-stable immonium salt that is an ionic liquid at moderately elevated temperature, useful in molten salt technique[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Fuller et al.
Journal of the Chemical Society. Chemical Communications, 299-299 (1994)
Capillary electrophoretic application of 1-alkyl-3-methylimidazolium-based ionic liquids.
Yanes EG, et al.
Analytical Chemistry, 73(16), 3838-3844 (2001)
Structure of 1-ethyl-3-methylimidazolium hexafluorophosphate: model for room temperature molten salts
aDe Long, Hugh C.
Journal of the Chemical Society. Chemical Communications, 3, 299-300 (1994)
Solubility of 1-alkyl-3-methylimidazolium hexafluorophosphate in hydrocarbons
Domanska, Urszula, and Andrzej Marciniak
Journal of Chemical and Engineering Data, 48.3, 451-456 (2003)
Jolanta Flieger et al.
Molecules (Basel, Switzerland), 20(12), 22058-22068 (2015-12-23)
A series of imidazolium and pyridinium ionic liquids with different anions (Cl(-), Br(-), BF₄(-), PF₆(-)) has been evaluated for their adsorption activity on silica gel. Quantification of the ionic liquids has been performed by the use of RP-HPLC with organic-aqueous

Articles

Functionalized imidazolium cations with thioether, urea, or thiourea derivatized side chains act as metal-ligating moieties, whereas the PF6– anions provide the desired water immiscibility.

Functionalized imidazolium cations with thioether, urea, or thiourea derivatized side chains act as metal-ligating moieties, whereas the PF6– anions provide the desired water immiscibility.

Functionalized imidazolium cations with thioether, urea, or thiourea derivatized side chains act as metal-ligating moieties, whereas the PF6– anions provide the desired water immiscibility.

Functionalized imidazolium cations with thioether, urea, or thiourea derivatized side chains act as metal-ligating moieties, whereas the PF6– anions provide the desired water immiscibility.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service