Skip to Content
Merck
All Photos(1)

Key Documents

94904

Sigma-Aldrich

Sodium 4-vinylbenzenesulfonate

technical, ≥90% (T)

Synonym(s):

4-Vinylbenzenesulfonic acid sodium salt, Styrene-4-sulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7NaO3S
CAS Number:
Molecular Weight:
206.19
Beilstein:
3575931
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

≥90% (T)

form

powder

mp

≥300 °C

functional group

sulfonic acid

SMILES string

[Na+].[O-]S(=O)(=O)c1ccc(C=C)cc1

InChI

1S/C8H8O3S.Na/c1-2-7-3-5-8(6-4-7)12(9,10)11;/h2-6H,1H2,(H,9,10,11);/q;+1/p-1

InChI key

XFTALRAZSCGSKN-UHFFFAOYSA-M

Related Categories

Application

Sodium 4-vinylbenzenesulfonate can be used as a starting material to synthesize poly(sodium 4-vinylbenzenesulfonate) via free radical and controlled radical polymerization.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinyi Guo et al.
Journal of nanobiotechnology, 17(1), 86-86 (2019-08-08)
Controllable and multiple DNA release is critical in modern gene-based therapies. Current approaches require complex assistant molecules for combined release. To overcome the restrictions on the materials and environment, a novel and versatile DNA release method using a nano-electromechanical (NEMS)
Graphene oxide as a radical initiator: Free radical and controlled radical polymerization of sodium 4-vinylbenzenesulfonate with graphene oxide
Voylov D, et al.
ACS Macro Letters, 5(2), 199-202 (2016)
Takuya Kubo et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 33(11), 1311-1315 (2017-11-14)
We report on a molecularly imprinted hydrogel for the selective fluorescent detection of a targeting protein. To achieve the fluorescent detection of proteins, a fluorescent functional monomer was synthesized with fluorescein isothiocyanate and allylamine. The monomer (FITC-AA) provided a gradable
Jiehua Lei et al.
ACS applied materials & interfaces, 9(34), 28209-28221 (2017-08-08)
Glycosaminoglycans (GAGs), especially heparin and heparan sulfate (HS), hold great potential for inducing the neural differentiation of embryonic stem cells (ESCs) and have brought new hope for the treatment of neurological diseases. However, the disadvantages of natural heparin/HS, such as
Shizhe Fu et al.
Nanoscale, 9(26), 9026-9033 (2017-06-24)
An efficient enzyme-powered micromotor device was fabricated by assembling multiple layers of catalase on the inner surface of a poly(3,4-ethylenedioxythiophene and sodium 4-styrenesulfonate)/Au microtube (PEDOT-PSS/Au). The catalase assembly was achieved by programmed DNA hybridization, which was performed by immobilizing a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service