Skip to Content
Merck
All Photos(1)

Key Documents

94077

Sigma-Aldrich

H-Pro-2-ClTrt resin

extent of labeling: ~0.6 mmol/g loading

Synonym(s):

L-Proline 2-chlorotrityl ester polymer-bound

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352116
NACRES:
NA.22

form

crystals

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

~0.6 mmol/g loading

particle size

75-150 μm

application(s)

peptide synthesis

storage temp.

2-8°C

Application

H-Pro-2-ClTrt resin is an acid-labile peptide reagent commonly used to prepare protected peptides by solid-phase peptide synthesis using the Fmoc strategy.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vitro and ex vivo studies on angiotensin-i converting enzyme (Ace) inhibitory activity of short synthetic peptides
Yakimova B, et al.
Farmacia, 69(2), 307-313 (2021)
Characterization of a new bioactive peptide from Potamotrygon gr. orbignyi freshwater stingray venom
Conceiccao K, et al.
Peptides, 30(12), 2191-2199 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service