932469
4,4′,4′′-Tris(N-(naphthalen-2-yl)-N-phenyl-amino)-triphenylamine
Synonym(s):
TSPO1, Diphenyl(4-(triphenylsilyl)phenyl)phosphine oxide
About This Item
Recommended Products
grade
sublimed grade
Quality Level
description
μh ≈ 1.1 x 10-6 cm2 V−1 s−1
μe ≈ 5.7 x 10-6 cm2 V−1 s−1
Assay
≥97% (HPLC)
loss
0.5% TGA, > 300 °C (weight loss)
mp
236 °C
solubility
chloroform: soluble
dichloromethane: soluble
toluene: soluble
λmax
266 nm±5 nm in dichloromethane
fluorescence
λem 298 nm±10 nm in dichloromethane
Orbital energy
HOMO 6.79 eV
LUMO 2.52 eV
Looking for similar products? Visit Product Comparison Guide
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service