Skip to Content
Merck
All Photos(1)

Key Documents

930148

Sigma-Aldrich

Glycerol carbonate-(carbonyl-13C)

≥99 atom % 13C, ≥99% (CP)

Synonym(s):

4-(Hydroxymethyl)-1,3-dioxolan-2-one-2-13C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC3H6O4
CAS Number:
Molecular Weight:
119.08
MDL number:
UNSPSC Code:
51113400
NACRES:
NA.13

isotopic purity

≥99 atom % 13C

Quality Level

Assay

≥99% (CP)

technique(s)

mass spectrometry (MS): suitable

mass shift

M+1

storage temp.

−20°C

SMILES string

OCC(O1)CO[13C]1=O

InChI

1S/C4H6O4/c5-1-3-2-7-4(6)8-3/h3,5H,1-2H2/i4+1

InChI key

JFMGYULNQJPJCY-AZXPZELESA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service