Skip to Content
Merck
All Photos(1)

Key Documents

454524

Sigma-Aldrich

Glycerol-1,1,2,3,3-d5

98 atom % D

Synonym(s):

1,2,3-Propanetriol-1,1,2,3,3-d5, Deuterated glycerol, Glycerin-1,1,2,3,3-d5

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCD2)2CDOH
CAS Number:
Molecular Weight:
97.12
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

technique(s)

NMR: suitable
bio NMR: suitable
protein expression: suitable

refractive index

n20/D 1.472 (lit.)

bp

182 °C (lit.)

mp

20 °C (lit.)

density

1.331 g/mL at 25 °C

mass shift

M+5

SMILES string

[2H]C([2H])(O)C([2H])(O)C([2H])([2H])O

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2/i1D2,2D2,3D

InChI key

PEDCQBHIVMGVHV-UXXIZXEISA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

320.0 °F - closed cup

Flash Point(C)

160 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xueqin Chen et al.
Journal of controlled release : official journal of the Controlled Release Society, 200, 78-86 (2015-01-01)
Understanding the penetration mechanisms of drugs into human skin is a key issue in pharmaceutical and cosmetics research. To date, the techniques available for percutaneous penetration of compounds fail to provide a quantitative 3D map of molecular concentration distribution in
Víctor A Lórenz-Fonfría et al.
Journal of the American Chemical Society, 137(5), 1850-1861 (2015-01-15)
Light-gated ion permeation by channelrhodopsin-2 (ChR2) relies on the photoisomerization of the retinal chromophore and the subsequent photocycle, leading to the formation (on-gating) and decay (off-gating) of the conductive state. Here, we have analyzed the photocycle of a fast-cycling ChR2
Emily Golden et al.
Analytical biochemistry, 485, 102-108 (2015-06-16)
Cholesterol oxidase (CO) is a FAD (flavin adenine dinucleotide) containing enzyme that catalyzes the oxidization and isomerization of cholesterol. Studies directed toward elucidating the catalytic mechanism of CO will provide an important general understanding of Flavin-assisted redox catalysis. Hydrogen atoms
Catalin Gainaru et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(49), 17402-17407 (2014-11-26)
We present the discovery of an unusually large isotope effect in the structural relaxation and the glass transition temperature Tg of water. Dielectric relaxation spectroscopy of low-density as well as of vapor-deposited amorphous water reveal Tg differences of 10 ±

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service