Skip to Content
Merck
All Photos(1)

Key Documents

924229

Sigma-Aldrich

(S,R,S)-AHPC-Me

95%

Synonym(s):

(2S,4R)-1-((S)-2-amino-3,3-dimethylbutanoyl)-4-hydroxy-N-((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)pyrrolidine-2-carboxamide, PROTAC® research ligand, VH032 methyl derivative

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H32N4O3S
CAS Number:
Molecular Weight:
444.59
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

ligand

VH032

Quality Level

Assay

95%

form

powder

storage temp.

2-8°C

SMILES string

C([C@H](C(C)(C)C)N)(=O)N1[C@H](C(N[C@@H](C)C2=CC=C(C=C2)C3=C(C)N=CS3)=O)C[C@@H](O)C1

InChI

1S/C23H32N4O3S/c1-13(15-6-8-16(9-7-15)19-14(2)25-12-31-19)26-21(29)18-10-17(28)11-27(18)22(30)20(24)23(3,4)5/h6-9,12-13,17-18,20,28H,10-11,24H2,1-5H3,(H,26,29)/t13-,17+,18-,20+/m0/s1

InChI key

JOSFQWNOUSNZBP-UUZHKXTQSA-N

Application

(S,R,S)-AHPC-Me is a VH032 derivative used in the recruitment of the von Hippel-Lindau (VHL) protein for targeted protein degradation and PROTAC (proteolysis-targeting chimeras) technology. This was previously listed under ATEH961B8E6E.

Related Technology Spotlight: Degrader Building Blocks for Targeted Protein Degradation

Browse our growing synthesis and research tools: Protein Degrader Building Blocks
Targeted protein degradation

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefan Vollmer et al.
Journal of medicinal chemistry, 63(1), 157-162 (2019-12-06)
PROteolysis TArgeting Chimeras (PROTACs) targeting the degradation of MEK have been designed based on allosteric MEK inhibitors. Inhibition of the phosphorylation of ERK1/2 was less effective with the PROTACs than a small-molecule inhibitor; the best PROTACs, however, were more effective
Jiantao Hu et al.
Journal of medicinal chemistry, 62(3), 1420-1442 (2019-04-17)
The estrogen receptor (ER) is a validated target for the treatment of estrogen receptor-positive (ER+) breast cancer. Here, we describe the design, synthesis, and extensive structure-activity relationship (SAR) studies of small-molecule ERα degraders based on the proteolysis targeting chimeras (PROTAC)
Marı A Maneiro et al.
ACS chemical biology, 15(6), 1306-1312 (2020-04-28)
Targeting protein degradation with Proteolysis-Targeting Chimeras (PROTACs) is an area of great current interest in drug discovery. Nevertheless, although the high effectiveness of PROTACs against a wide variety of targets has been established, most degraders reported to date display limited
Cyrille S Kounde et al.
Chemical communications (Cambridge, England), 56(41), 5532-5535 (2020-04-17)
With the intent of achieving greater spatiotemporal control of PROTAC-induced protein degradation, a light-activated degrader was designed by photocaging an essential E3 ligase binding motif in a BRD4 targeting PROTAC. Proteolysis was triggered only after a short irradiation time, the
Mingliang Wang et al.
Journal of medicinal chemistry, 63(14), 7510-7528 (2020-05-22)
Src homology 2 domain-containing phosphatase 2 (SHP2) is an attractive therapeutic target for human cancers and other human diseases. Herein, we report our discovery of potent small-molecule SHP2 degraders whose design is based upon the proteolysis-targeting chimera (PROTAC) concept. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service