Skip to Content
Merck
All Photos(1)

Key Documents

921025

Sigma-Aldrich

Palladium(II) acetylacetonate

≥99.9% trace metals basis

Synonym(s):

Palladium(II) 2,4-pentanedionate, Pd(acac)2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd(C5H7O2)2
CAS Number:
Molecular Weight:
304.64
Beilstein:
4136188
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.23
Assay:
≥99.9% trace metals basis

Quality Level

Assay

≥99.9% trace metals basis

mp

200-251 °C (dec.)

SMILES string

CC(=O)\C=C(\C)O[Pd]O\C(C)=C/C(C)=O

InChI

1S/2C5H8O2.Pd/c2*1-4(6)3-5(2)7;/h2*3,6H,1-2H3;/q;;+2/p-2/b2*4-3-;

InChI key

JKDRQYIYVJVOPF-FDGPNNRMSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Palladium(II) 2,4-pentanedionate can be used to synthesize palladium(II) complexes applicable in common cross-coupling reactions like Suzuki, Heck and Buchwald-Hartwig coupling. As a palladium precursor, it can be used to synthesize hollow Pd/Pt single-crystalline nanocubes with enhanced electrocatalytic activity due to increased surface area. The surface modification of TiO2 using Pd(acac)2 as a metal-organic precursor has enhanced its photocatalytic activity under UV irradiation. Pd(acac)2 can also be used to develop bimetallic alloy NP catalysts applicable in formic acid dehydrogenation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New insights into catalysis for Heck reactions with fine supported Pd particles.
Huang L, et al.
Reaction Chemistry & Engineering, 5(5), 921-934 (2020)
Madhu Bala et al.
Turkish journal of biology = Turk biyoloji dergisi, 43, 154-154 (2019-08-15)
[This corrects the article DOI: 10.3906/biy-1412-46.].
Xiaoqing Huang et al.
Angewandte Chemie (International ed. in English), 48(26), 4808-4812 (2009-05-16)
Efficiency simplified: A synthetic strategy has been developed to prepare single-crystalline hollow Pd/Pt nanocubes (right, see picture; left: nanocubes). Compared to the solid Pd/Pt nanocubes of similar sizes, the hollow Pd/Pt nanocubes increase accessible surface area and therefore improve electrocatalytic
Deposition of Pd nanoparticles on TiO2 using a Pd (acac)2 precursor for photocatalytic oxidation of CO under UV-LED irradiation.
Selishchev D S, et al.
Applied Catalysis. B, Environmental, 235, 214-224 (2018)
Önder Metin et al.
Nanoscale, 5(3), 910-912 (2012-12-21)
Monodisperse 4 nm AuPd alloy nanoparticles with controlled composition were synthesized by co-reduction of hydrogen tetrachloroaurate(III) hydrate and palladium(II) acetylacetonate with a borane-morpholine complex in oleylamine. These NPs showed high activity (TOF = 230 h(-1)) and stability in catalyzing formic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service