Skip to Content
Merck
All Photos(3)

Key Documents

911992

Sigma-Aldrich

TBSAB

≥95%

Synonym(s):

Tributylsulfoammonium betaine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H27NO3S
CAS Number:
Molecular Weight:
265.41
UNSPSC Code:
12352200
NACRES:
NA.22

Assay

≥95%

form

powder or crystals

reaction suitability

reagent type: oxidant

mp

94-99 °C

functional group

amine

Application

TBSAB is a sulfation reagent that efficiently converts alcohols to organosulfates under mild conditions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel M Gill et al.
Chemical communications (Cambridge, England), 55(30), 4319-4322 (2019-03-19)
The study of organosulfates is a burgeoning area in biology, yet there are significant challenges with their synthesis. We report the development of a tributylsulfoammonium betaine as a high yielding route to organosulfates. The optimised reaction conditions were interrogated with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service