Skip to Content
Merck
All Photos(1)

Key Documents

373680

Sigma-Aldrich

Sulfur trioxide N,N-dimethylformamide complex

97%

Synonym(s):

DMF-sulfur trioxide complex

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCON(CH3)2 · SO3
CAS Number:
Molecular Weight:
153.16
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

reaction suitability

reagent type: oxidant

mp

155-158 °C (lit.)

storage temp.

2-8°C

SMILES string

O=S(=O)=O.[H]C(=O)N(C)C

InChI

1S/C3H7NO.O3S/c1-4(2)3-5;1-4(2)3/h3H,1-2H3;

InChI key

AFDQGRURHDVABZ-UHFFFAOYSA-N

Application

Reactant for preparation of:
  • Aminopyridines as inhibitors of tetrameric fructose-1,6-bisphosphatase
  • Cyclic sulfates
  • A dual acid site-grafted zirconium hydroxide-anchored catalyst
Widely used mild sulfating agent.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Journal of the Chemical Society. Perkin Transactions 1, 1739-1739 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service