Skip to Content
Merck
All Photos(1)

Key Documents

911895

D-Sorbitol-1,1,2,3,4,5,6,6-d8

98 atom % D, ≥99% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D8H6O6
CAS Number:
Molecular Weight:
190.22
UNSPSC Code:
12142201

isotopic purity

98 atom % D

Assay

≥99% (CP)

form

solid

SMILES string

O[C@@]([C@@]([C@]([C@](C([2H])(O)[2H])([2H])O)([2H])O)([2H])O)([2H])C([2H])(O)[2H]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service