Skip to Content
Merck
All Photos(1)

Key Documents

605514

Sigma-Aldrich

D-Sorbitol-13C6

99 atom % 13C, 99% (CP)

Synonym(s):

D-Glucitol-13C6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6H14O6
Molecular Weight:
188.13
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

99% (CP)

form

solid

optical activity

[α]25/D +101°, c = 0.5 in acidified ammonium molybdate

mp

98-100 °C (lit.)

mass shift

M+6

SMILES string

O[13CH2][13C@@H](O)[13C@@H](O)[13C@H](O)[13C@@H](O)[13CH2]O

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4+,5-,6-/m1/s1/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

FBPFZTCFMRRESA-GVZKVBLJSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali Kiani-Pouya et al.
Plant, cell & environment, 40(9), 1900-1915 (2017-05-31)
Epidermal bladder cells (EBCs) have been postulated to assist halophytes in coping with saline environments. However, little direct supporting evidence is available. Here, Chenopodium quinoa plants were grown under saline conditions for 5 weeks. One day prior to salinity treatment, EBCs
Christian Scherling et al.
PloS one, 5(9), e12569-e12569 (2010-09-11)
In spite of evidence for positive diversity-productivity relationships increasing plant diversity has highly variable effects on the performance of individual plant species, but the mechanisms behind these differential responses are far from being understood. To gain deeper insights into the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service