Skip to Content
Merck
All Photos(1)

Key Documents

907782

Sigma-Aldrich

NanoFabTx reagent

PLGA-Nano

Synonym(s):

PLGA-Nano

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.25

Quality Level

form

pellets

application(s)

advanced drug delivery

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

NanoFabTx reagent, PLGA-Nano is a component of our NanoFabTx drug formulation kits (909637, 917737), which include additional reagents and protocols for nanoprecipitation and microfluidics to synthesize 100-200 nm nanoparticles. NanoFabTx reagent, PLGA-Nano is a poly(lactic-co-glycolic acid) (PLGA) polymer. PLGA is a biocompatible and biodegradable polymer that has been used widely in drug delivery systems for controlled drug release of many different types of therapeutic molecules.

Application

NanoFabTx reagent, PEGPLGA-50H can be used as a refill for the component in our NanoFabTx drug formulation kits (909637,

Legal Information

NANOFABTX is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service