900289
Poly(L-lactide-co-glycolide)
lactide:glycolide 20:80, viscosity 1.6 dL/g
Synonym(s):
PLGA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(C2H2O2)m(C3H4O2)nCH3
UNSPSC Code:
12162002
NACRES:
NA.23
Recommended Products
form
chips or granules
Quality Level
feed ratio
lactide:glycolide 20:80
viscosity
1.6 dL/g
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Cristiane R Oliveira et al.
Chemosphere, 236, 124282-124282 (2019-07-20)
Nanotechnology has the potential to overcome the challenges of sustainable agriculture, and nanopesticides can control agricultural pests and increase farm productivity with little environmental impact. However, it is important to evaluate their toxicity on non-target organisms, such as honeybees (Apis
Articles
Innovations in polymer technology have had a significant impact on the advancement of novel drug delivery systems.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service