Skip to Content
Merck
All Photos(1)

Key Documents

906522

Sigma-Aldrich

SLAM-Iγ2-13CH3 Methyl Labeling Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.12

technique(s)

bio NMR: suitable

Quality Level

storage temp.

−70°C

Related Categories

General description

SLAM-Iγ2-13CH3 Kit has 13CH3 isotopomer precursor and contains protocol instructions for creation of isotopically-labeled proteins.

Application

For detection of long-range nOes or for the NMR study of large proteins
SLAM-Iγ2-13CH3 is available for labeling isoleucine precursor with 13CH3 isotopomer. This kit has been tested with protein isotopic labeling in E. coli. It enables study of protein complexes revealing the mechanisms of dynamic molecular machines in solid-state isotopomer.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zachary K Boswell et al.
Biochemistry, 58(3), 144-155 (2018-10-20)
NMR spectroscopy is particularly adept at site-specifically monitoring dynamic processes in proteins, such as protein folding, domain movements, ligand binding, and side-chain rotations. By coupling the favorable spectroscopic properties of highly dynamic side-chain methyl groups with transverse-relaxation-optimized spectroscopy (TROSY), it
Stereospecific isotopic labeling of methyl groups for NMR spectroscopic studies of high-molecular-weight proteins.
Pierre Gans et al.
Angewandte Chemie (International ed. in English), 49(11), 1958-1962 (2010-02-17)
Silke Wiesner et al.
Current opinion in structural biology, 35, 60-67 (2015-09-26)
Intermolecular interactions are indispensible for biological function. Here we discuss how novel NMR techniques can provide unique insights into the assembly, dynamics and regulation of biomolecular complexes. We focus on applications that exploit the methyl TROSY effect and show that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service