All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H13BrO4
CAS Number:
Molecular Weight:
241.08
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22
Recommended Products
Assay
≥95%
form
liquid
reaction suitability
reagent type: linker
functional group
bromo
carboxylic acid
storage temp.
−20°C
SMILES string
BrCCOCCOCCC(O)=O
InChI key
CCEBKJLCKKPYHB-UHFFFAOYSA-N
Related Categories
Application
Bromo-PEG2-acid is a heterobifunctional, PEGylated crosslinker featuring a bromo group at one end and a carboxyl group at the other. The hydrophillic PEG linker facilitates solubility in biological applications. Bromo-PEG2-acid can be used for bioconjugation or as a building block for synthesis of small molecules, conjugates of small molecules and/or biomolecules, or other tool compounds for chemical biology and medicinal chemistry that require ligation. Examples of applications include its synthetic incorporation into antibody-drug conjugates or proteolysis-targeting chimeras (PROTAC® molecules) for targeted protein degradation.
This is a crosslinker with a bromo group on one end and a carboxyl group on the other end. The compound contains two PEG units to help improve solubility.
Legal Information
PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license
related product
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service