Skip to Content
Merck
All Photos(1)

Key Documents

14625

Sigma-Aldrich

Bis[2-(2-chloroethoxy)ethyl] ether

≥99.0% (T)

Synonym(s):

1,11-Dichloro-3,6,9-trioxaundecane, Tetraethylene glycol dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O(CH2CH2OCH2CH2Cl)2
CAS Number:
Molecular Weight:
231.12
Beilstein:
1071710
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0% (T)

refractive index

n20/D 1.464

density

1.18 g/mL at 20 °C (lit.)

functional group

chloro
ether

SMILES string

ClCCOCCOCCOCCCl

InChI

1S/C8H16Cl2O3/c9-1-3-11-5-7-13-8-6-12-4-2-10/h1-8H2

InChI key

ZCFRYTWBXNQVOW-UHFFFAOYSA-N

Application

Bis[2-(2-chloroethoxy)ethyl] ether was used in preparation of series of para-(1,1,3,3-tetramethylbutyl)-phenoxypoly(ethoxy)ethanols. It was used in the synthesis of 1-(4-pyridiniumaldoxime)-2-(((2-chloroethoxy)-2-ethoxy)ethoxy)ethane.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Hawrył et al.
Molecules (Basel, Switzerland), 25(18) (2020-09-24)
The aim of this study was to evaluate the ability of multivariate techniques to predict antioxidant and cytotoxic activity of the selected lichens from the chromatographic data. A simple and reproducible HPLC-DAD technique has been used to obtain the chromatographic
The preparation of a series of molecularly homogeneous para-t-octylphenoxypoly (ethoxy) ethanols.
Mansfield RC and Locke JE.
Journal of the American Oil Chemists' Society, 41(4), 267-272 (1964)
M C de Koning et al.
Bioorganic & medicinal chemistry, 19(1), 588-594 (2010-11-30)
A conceptually novel approach to the design of reactivators of nerve agent-inhibited acetylcholinesterase (AChE) is presented. The concept comprises the linkage of a peripheral site ligand via a spacer to a reactivating moiety with the eventual goal to develop non-ionic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service