Skip to Content
Merck
All Photos(1)

Key Documents

901410

Sigma-Aldrich

Hexagonal boron nitride ink

greener alternative

for inkjet printing

Synonym(s):

Hexagonal boron nitride dispersion, with ethyl cellulose in cyclohexanone and terpineol, hBN dispersion, hBN inkjet printable ink

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BN
CAS Number:
Molecular Weight:
24.82
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

form

dispersion

composition

Solid content, 4.4-6.4%

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

white

particle size

≤0.3 μm

surface tension

30-35 dyn/cm

viscosity

6-15 cP (shear viscosity at 1000 s-1, 25 °C)

bp

213-218 °C (Terpineol)

density

0.9-1.1 g/mL at 25 °C

greener alternative category

SMILES string

B#N

InChI

1S/BN/c1-2

InChI key

PZNSFCLAULLKQX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Curing condition: 250-350 °C, 20-30 min.
Hexagonal boron nitride ink (hBN) is an insulating material with properties like high thermal conductivity, excellent dimensional stability, and good mechanical robustness. hBN inks can be used in the fabrication of flexible memory devices.[1][2][3]
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Boron nitride materials have interesting properties like large energy band gap, good resistance to oxidation, excellent thermal stability, thermal conductivity, chemical inertness, significance mechanical property and widespred applications. Click here for more information.

Application

This ink was tested on polyimide and silicon dioxide substrates.

Legal Information

Sold under Materials Transfer Agreement with Mark Hersam group at Northwestern University.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

115.7 °F

Flash Point(C)

46.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2D nanocomposite of hexagonal boron nitride nanoflakes and molybdenum disulfide quantum dots applied as the functional layer of all-printed flexible memory device
Rehman MM, et al.
Materials Research Bulletin, 105(6), 28-35 (2018)
Screen-printable electronic ink of ultrathin boron nitride nanosheets
Joseph AM, et al.
ACS Omega, 1(6), 1220-1228 (2016)
Large scale growth and characterization of atomic hexagonal boron nitride layers
Song L, et al.
Nano Letters, 10(8), 3209-3215 (2010)

Articles

Recent advancements in paper-based sensing platforms offer cost-effective clinical diagnostics with microfluidic channels and colorimetric or electrochemical detection zones.

Recent advancements in paper-based sensing platforms offer cost-effective clinical diagnostics with microfluidic channels and colorimetric or electrochemical detection zones.

Recent advancements in paper-based sensing platforms offer cost-effective clinical diagnostics with microfluidic channels and colorimetric or electrochemical detection zones.

Recent advancements in paper-based sensing platforms offer cost-effective clinical diagnostics with microfluidic channels and colorimetric or electrochemical detection zones.

See All

Questions

  1. What is the size and morphology of the particles?

    1 answer
    1. The particle size is specified to be ≤0.3 μm, but there is no other available information regarding morphology.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service