Skip to Content
Merck
All Photos(1)

Key Documents

901867

Sigma-Aldrich

Molybdenum disulfide ink for spin/spray coating

greener alternative

Synonym(s):

MoS2 dispersion, MoS2 ink

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MoS2
UNSPSC Code:
12352103
NACRES:
NA.23

description

Resistivity: 75-100 kΩ·cm; Prepared by spin-coating at 1000 rpm/30 s for 1 coat, followed by thermal annealing at 400°C in Ar/H2 for 3 hours

form

dispersion

composition

Solid Content, 1.5-2.5%

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

green

particle size

≤0.4 μm

viscosity

2-4 mPa.s

bp

78 °C (Ethanol)

density

0.78-0.88 g/mL at 25 °C

greener alternative category

SMILES string

[Mo]=S=S

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Click here for more information.

Application

n-type semiconductor for use in transistors, photodetectors and photovoltaics

Legal Information

Sold under Materials Transfer Agreement with Mark Hersam group at Northwestern University

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deep Jariwala et al.
Nano letters, 16(1), 497-503 (2015-12-15)
The recent emergence of a wide variety of two-dimensional (2D) materials has created new opportunities for device concepts and applications. In particular, the availability of semiconducting transition metal dichalcogenides, in addition to semimetallic graphene and insulating boron nitride, has enabled

Articles

Recent advancements in paper-based sensing platforms offer cost-effective clinical diagnostics with microfluidic channels and colorimetric or electrochemical detection zones.

Recent advancements in paper-based sensing platforms offer cost-effective clinical diagnostics with microfluidic channels and colorimetric or electrochemical detection zones.

Recent advancements in paper-based sensing platforms offer cost-effective clinical diagnostics with microfluidic channels and colorimetric or electrochemical detection zones.

Recent advancements in paper-based sensing platforms offer cost-effective clinical diagnostics with microfluidic channels and colorimetric or electrochemical detection zones.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service