Skip to Content
Merck
All Photos(1)

Key Documents

901255

Sigma-Aldrich

1-Ethoxy-2-propanol

≥95%

Synonym(s):

1-Ethoxypropan-2-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12O2
CAS Number:
Molecular Weight:
104.15
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

Quality Level

Assay

≥95%

form

liquid

refractive index

n/D 1.407

density

0.898 g/mL

functional group

ether
hydroxyl

storage temp.

2-8°C

InChI

1S/C5H12O2/c1-3-7-4-5(2)6/h5-6H,3-4H2,1-2H3

InChI key

JOLQKTGDSGKSKJ-UHFFFAOYSA-N

Application

Useful building block for synthesis. 1-Ethoxy-2-propanol may be used in the preparation of 1-ethoxy-2-bromopropane using phosphorus tribromide. This product was previously listed as CDS000221.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

101.3 °F

Flash Point(C)

38.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reaction of 2-Alkoxymethylthiiranes with Lithium Aluminum Hydride and with Secondary Amines.
Jacob R L, et al.
The Journal of Organic Chemistry, 26 (9), 3472-3476 (1961)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service