Skip to Content
Merck
All Photos(1)

Key Documents

38457

Sigma-Aldrich

3-Methoxy-1-propanol

≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H10O2
CAS Number:
Molecular Weight:
90.12
Beilstein:
1731204
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (GC)

refractive index

n20/D 1.413

bp

150-152 °C

density

0.942 g/mL at 20 °C (lit.)

functional group

ether
hydroxyl

SMILES string

COCCCO

InChI

1S/C4H10O2/c1-6-4-2-3-5/h5H,2-4H2,1H3

InChI key

JDFDHBSESGTDAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methoxy-1-propanol is a primary alcohol. Kinetics of the reactions of 3-methoxy-1-propanol with phenyl isocyanate in the presence of metal catalysts has been reported. Selective synthesis of 3-methoxy-1-propanol from methanol and allyl alcohol with metal oxides and zeolites catalysts in the liquid phase has been studied.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective synthesis of 3-methoxy-1-propanol from methanol and allyl alcohol with metal oxide catalysts.
Yamakawa T, et al.
Catalysis Communications, 2(6), 191-194 (2001)
Kinetics of alcohol-isocyanate reactions with metal catalysts.
Rand L, et al.
Journal of Applied Polymer Science, 9(5), 1787-1795 (1965)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service