Skip to Content
Merck
All Photos(2)

Key Documents

901183

Sigma-Aldrich

Propylhydrazine hydrochloride

Synonym(s):

1-Propylhydrazine HCl, Nucleophilic hydrazine probe

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H10N2 · xHCl
CAS Number:
Molecular Weight:
74.12 (free base basis)
MDL number:
UNSPSC Code:
12352101

form

solid

availability

available only in USA

storage temp.

2-8°C

SMILES string

[H]Cl.NNCCC

Application

Propylhydrazine can be used as synthetic building blocks for various applications. Ben Cravatt′s lab recently developed clickable, nucleophilic hydrazine probes for "reverse-polarity" activity-based protein profiling, or (RP)-ABPP, where this propylhydrazine was used as a competitor for hydrazine probe 901210.

related product

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service