699101
1-Boc-1-methylhydrazine
97%
Synonym(s):
2-Methylcarbazic acid tert-butyl ester, N-(tert-Butoxycarbonyl)-N-methylhydrazine, N-Boc-N-methylhydrazine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C6H14N2O2
CAS Number:
Molecular Weight:
146.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
liquid
refractive index
n20/D 1.436
density
0.985 g/mL at 25 °C
functional group
amine
hydrazide
SMILES string
CN(N)C(=O)OC(C)(C)C
InChI
1S/C6H14N2O2/c1-6(2,3)10-5(9)8(4)7/h7H2,1-4H3
InChI key
IHMQNZFRFVYNDS-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
160.0 °F
Flash Point(C)
71.1 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service