Skip to Content
Merck
All Photos(1)

Key Documents

719870

Sigma-Aldrich

Resomer® RG 503 H, Poly(D,L-lactide-co-glycolide)

acid terminated, lactide:glycolide 50:50, Mw 24,000-38,000

Synonym(s):

Poly(D,L-lactide-co-glycolide), PLGA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C3H4O2]x[C2H2O2]y
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

Quality Level

form

amorphous

feed ratio

lactide:glycolide 50:50

mol wt

Mw 24,000-38,000

degradation timeframe

<3 months

viscosity

0.32-0.44 dL/g, 0.1 % (w/v) in chloroform(25 °C, Ubbelohde) (size 0c glass capillary viscometer)

transition temp

Tg 44-48 °C

storage temp.

2-8°C

SMILES string

OCC(O)=O.CC(O)C(O)=O

Looking for similar products? Visit Product Comparison Guide

Application

Controlled release

Features and Benefits

Controlled release of bioactive agents, sutures and bioabsorbable implantable devices.

Physical form

Biocompatible, biodegradable polymer.

Legal Information

Product of Evonik
RESOMER is a registered trademark of Evonik Rohm GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaamini M Dhanabalan et al.
Biomaterials science, 8(15), 4308-4321 (2020-07-01)
Osteoarthritis (OA) is a joint disease characterized by progressive damage of articular cartilage and the adjoining subchondral bone. Chondrocytes, the primary cells of the cartilage, have limited regenerative capacity and when they undergo stress due to trauma or with aging
Rongcai Liang et al.
International journal of pharmaceutics, 454(1), 344-353 (2013-07-23)
Peptide or protein degradation often occurs when water flows into the dosage form. The aim of this study was to investigate the effect of water on exenatide acylation in poly(lactide-co-glycolide) (PLGA) microspheres. Exenatide-loaded PLGA microspheres were incubated at different relative
Vanna Sanna et al.
International journal of nanomedicine, 7, 5501-5516 (2012-10-25)
Resveratrol, like other natural polyphenols, is an extremely photosensitive compound with low chemical stability, which limits the therapeutic application of its beneficial effects. The development of innovative formulation strategies, able to overcome physicochemical and pharmacokinetic limitations of this compound, may
Shu-Chun Chuang et al.
Parasites & vectors, 6, 34-34 (2013-02-13)
Current development efforts of subunit vaccines against Toxoplasma gondii, the etiological agent of toxoplasmosis, have been focused mainly on tachyzoite surface antigen 1 (SAG1). Since microparticles made from poly (lactide-co-glycolide) (PLG) polymers have been developed as safe, potent adjuvants or
Mani Gajendiran et al.
Colloids and surfaces. B, Biointerfaces, 104, 107-115 (2013-01-10)
A series of biodegradable low molecular weight PLGA-PEG-PLGA tri-block copolymers have been synthesized in powder form. The anti-tuberculosis drug Isoniazid (INH) loaded polymeric core-shell nanoparticles (CSNPs) have been prepared by sonication followed by water-in-oil-in-water (w/o/w) double emulsification technique. The nanoparticles

Articles

Interest in utilizing biodegradable polymers for biomedical applications has grown since the 1960s.

Synthetic aliphatic polyesters dominate resorbable biomaterials in clinical use.

Synthetic aliphatic polyesters dominate resorbable biomaterials in clinical use.

Synthetic aliphatic polyesters dominate resorbable biomaterials in clinical use.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service