Skip to Content
Merck
All Photos(1)

Key Documents

86778

Sigma-Aldrich

3′,3″,5′,5″-Tetrabromophenolphthalein ethyl ester

for microscopy

Synonym(s):

TBPE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14Br4O4
CAS Number:
Molecular Weight:
661.96
Beilstein:
2228548
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

for microscopy

Quality Level

form

solid

SMILES string

CCOC(=O)c1ccccc1\C(c2cc(Br)c(O)c(Br)c2)=C3\C=C(Br)C(=O)C(Br)=C3

InChI

1S/C22H14Br4O4/c1-2-30-22(29)14-6-4-3-5-13(14)19(11-7-15(23)20(27)16(24)8-11)12-9-17(25)21(28)18(26)10-12/h3-10,27H,2H2,1H3

InChI key

SQFXATUXPUCFFO-UHFFFAOYSA-N

Application

3′,3″,5′,5″-Tetrabromophenolphthalein ethyl ester (TBPE) is an anionic dye used as a counter ion in the spectrophotometric determination of cationic surfactants. TBPE is also used in the spectrometric determination of aliphatic primary, secondary and tertiary amines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous spectrophotometric analysis of aliphatic amines utilizing thermochromism of charge-transfer complexes with tetrabromophenolphthalein ethyl ester.
Sakai T, et al.
Analytical Sciences, 21(12), 1557-1560 (2005)
Simple method for spectrophotometric determination of cationic surfactants without liquid-liquid extraction.
Kamaya M, et al.
Analytica Chimica Acta, 384(2), 215-218 (1999)
H T Chang et al.
Electrophoresis, 22(3), 394-398 (2001-03-22)
Electroelution of protein bands from a gel has advantages over the competitive common technique requiring gel sectioning with respect to yield, speed and the potential for computer-controlled application to multicomponent two-dimensional (2-D) gels. The electroelution design for the commercial high-performance
[Pharmaco-analytical application of tetrabromophenolphthalein ethyl ester indicator dye. III. Interaction between alkaloids and TBPE].
G Szász et al.
Acta pharmaceutica Hungarica, 50(4), 161-165 (1980-07-01)
Ion association titration for the determination of local anesthetics in pharmaceuticals with tetrabromophenolphthalein ethyl ester as an indicator.
T Sakai et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 17(9), 1105-1107 (2001-11-16)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service