Skip to Content
Merck
All Photos(1)

Key Documents

85356

Sigma-Aldrich

Silicon dioxide

granular, ≥99.9%

Synonym(s):

Cristobalite, Quartz, Sand, Silica

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

≥99.9%

form

granular

manufacturer/tradename

Umicore Materials AG

refractive index

n20/D 1.544 (lit.)

particle size

0.2-0.7 mm

mp

1610 °C (lit.)

density

2.6 g/mL at 25 °C (lit.)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

SiO2 is majorly used as a substrate material with excellent thermo-mechanical properties in a variety of applications which include: vapour deposition[1][2], phase deposition[3], atomic force microscopy probes(AFM[4], spin coating[5], and electronic based devices[6].

Application

  • Bioresorbable pressure sensors protected with thermally grown silicon dioxide for the monitoring of chronic diseases and healing processes: Discusses the development of bioresorbable pressure sensors using silicon dioxide, highlighting its potential in medical diagnostics and monitoring (J Shin et al., 2019, Nature).
  • Solvent-assisted lipid bilayer formation on silicon dioxide and gold: This research investigates the interaction between lipid bilayers and silicon dioxide surfaces, useful in the development of biosensors and other biotechnological applications (SR Tabaei et al., 2014, ACS Publications).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Measurement of the adhesion force between carbon nanotubes and a silicon dioxide substrate.
Whittaker J D, et al.
Nano Letters, 6(5), 953-957 (2006)
Exciton diffusion measurements in poly (3-hexylthiophene).
Shaw P E, et al.
Advanced Materials, 20(18), 3516-3520 (2008)
Growth model of bamboo-shaped carbon nanotubes by thermal chemical vapor deposition.
Lee C J, et al.
Applied Physics Letters, 77(21), 3397-3399 (2000)
The effect of calcination temperature on the surface microstructure and photocatalytic activity of TiO2 thin films prepared by liquid phase deposition.
Yu J, et al
The Journal of Physical Chemistry B, 107(50), 13871-13879 (2003)
Oxygen-aided synthesis of polycrystalline graphene on silicon dioxide substrates.
Chen J, et al.
Journal of the American Chemical Society, 133(44), 17548-17551 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service