Skip to Content
Merck
All Photos(2)

Key Documents

860581

Sigma-Aldrich

α-Methyl-α-phenylsuccinimide

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11NO2
CAS Number:
Molecular Weight:
189.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

83-85 °C (lit.)

functional group

phenyl

SMILES string

CC1(CC(=O)NC1=O)c2ccccc2

InChI

1S/C11H11NO2/c1-11(7-9(13)12-10(11)14)8-5-3-2-4-6-8/h2-6H,7H2,1H3,(H,12,13,14)

InChI key

UDESUGJZUFALAM-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmacokinetics of N-desmethylmethsuximide in pediatric patients.
M V Miles et al.
The Journal of pediatrics, 114(4 Pt 1), 647-650 (1989-04-01)
N Wad et al.
Clinical neuropharmacology, 22(4), 239-240 (1999-08-12)
Serum protein binding of desmethyl-methsuximide (DM-MSM) in serum from 23 patients on polytherapy were determined using ultrafiltration and high-performance liquid chromatography. Desmethyl-methsuximide, The active metabolite of methsuximide (MSM), was found to have a moderate protein binding ranging between 45% and
D S Cardella et al.
Clinical biochemistry, 21(6), 329-331 (1988-12-01)
A method for the determination of the anti-epileptic drug methsuximide (MSM) and its active metabolite N-desmethylmethsuximide (NDM) is presented. 5-Methyl-5-phenylhydantoin is used as the internal standard. A simple solid-phase extraction procedure utilizing disposable reversed-phase C18 columns is described. Samples are
G K Szabo et al.
Clinical chemistry, 28(1), 100-104 (1982-01-01)
We describe an improved "high-pressure" liquid-chromatographic assay for simultaneous determination in serum of the five major antiepileptic drugs (ethosuximide, primidone, phenobarbital, phenytoin, and carbamazepine) and N-desmethylmethsuximide (the compound that must be quantitated for therapeutic drug monitoring of the antiepileptic drug
D A Coulter et al.
British journal of pharmacology, 100(4), 800-806 (1990-08-01)
1. Succinimide derivatives can be either convulsant (tetramethylsuccinimide (TMS)), or anticonvulsant (ethosuximide (ES); alpha-methyl-alpha-phenylsuccinimide (MPS)). ES, an anticonvulsant succinimide, has previously been shown to block calcium currents of thalamic neurones, while the convulsant succinimide TMS blocks gamma-aminobutyric acid (GABA) responses

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service