Skip to Content
Merck
All Photos(2)

Key Documents

851299

Sigma-Aldrich

4-Nitro-DL-phenylalanine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
210.19
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

mp

236-237 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

NC(Cc1ccc(cc1)[N+]([O-])=O)C(O)=O

InChI

1S/C9H10N2O4/c10-8(9(12)13)5-6-1-3-7(4-2-6)11(14)15/h1-4,8H,5,10H2,(H,12,13)

InChI key

GTVVZTAFGPQSPC-UHFFFAOYSA-N

Application

4-Nitro-DL-phenylalanine may be used as an internal standard for the determination of β-N-methylamino-L-alanine (L-BMAA) in environmental aqueous samples using proton nuclear magnetic resonance (1H NMR) technique.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Daniel Sojka et al.
Parasites & vectors, 1(1), 7-7 (2008-03-20)
Ticks are vectors for a variety of viral, bacterial and parasitic diseases in human and domestic animals. To survive and reproduce ticks feed on host blood, yet our understanding of the intestinal proteolytic machinery used to derive absorbable nutrients from
Li-Na Qu et al.
Hybridoma and hybridomics, 22(6), 401-406 (2003-12-20)
3-Nitro-L-tyrosine (nitrotyrosine) has recently been considered to be useful as a biomarker of endogenous production of several reactive nitrogen species including peroxynitrite. In the present study, nitrotyrosine was coupled to human serum albumin (HSA) using a two-step glutaraldehyde method and
L Morandeau et al.
Bioconjugate chemistry, 16(1), 184-193 (2005-01-20)
The feasibility of two-step radioimmunotherapy (RIT) of cancer by the Affinity Enhancement System (AES) has been demonstrated in experimental and clinical studies. This technique, associating a bispecific antibody and a bivalent peptide radiolabeled with iodine-131, has been developed to reduce
P W Schiller et al.
Life sciences, 33 Suppl 1, 319-322 (1983-01-01)
For the purpose of comparing the structural requirements of opioid receptor subsites interacting with phenylalanine residues of opioid peptides, analogs containing p-nitrophenylalanine (Phe(pNO2) ) were synthesized and tested in the guinea pig ileum (GPI) and mouse vas deferens (MVD) assay
A G Peranteau et al.
Analytical biochemistry, 227(1), 242-245 (1995-05-01)
The intrinsic fluorescence of tyrosine increases by a factor of approximately two when the carboxy group is liberated from a peptide bond by hydrolysis. The increase in fluorescence provides a novel way to monitor the hydrolysis of native tyrosine peptides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service