Skip to Content
Merck
All Photos(3)

Key Documents

104655

Sigma-Aldrich

1,1′-Bi-2-naphthol

99%

Synonym(s):

(±)-1,1′-Binaphthalene-2,2′-diol, (±)-2,2′-Dihydroxy-1,1′-dinaphthyl, 2,2′-Dihydroxybinaphthalene, 2,2′-Dihydroxydinaphthyl, 2,2′-Dinaphthol, BINOL

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC10H6C10H6OH
CAS Number:
Molecular Weight:
286.32
Beilstein:
997518
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

reaction suitability

reagent type: ligand

mp

214-217 °C (lit.)

SMILES string

Oc1ccc2ccccc2c1-c3c(O)ccc4ccccc34

InChI

1S/C20H14O2/c21-17-11-9-13-5-1-3-7-15(13)19(17)20-16-8-4-2-6-14(16)10-12-18(20)22/h1-12,21-22H

InChI key

PPTXVXKCQZKFBN-UHFFFAOYSA-N

Application

Chiral ligand and auxiliary for asymmetric Michael addition reaction; enantioselective Diels-Alder reaction; alkynylation.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yifeng Zhou et al.
Organic letters, 6(23), 4147-4149 (2004-11-05)
The readily available and inexpensive (S)-BINOL ligand in combination with Ti(O(i)Pr)(4) is an effective chiral catalyst for the catalytic asymmetric addition of alkynylzinc to unactivated simple ketones. Good to excellent enantioselectivities were achieved. No previous case has been reported successfully
Na Ji et al.
Journal of the American Chemical Society, 128(27), 8845-8848 (2006-07-06)
Chiral sum-frequency (SF) spectroscopy that measures both the real and the imaginary components of the SF spectral response was demonstrated for the first time. It was based on interference of the SF signal with a dispersionless SF reference. Solutions of
Yolanda Pérez-Fuertes et al.
Nature protocols, 3(2), 210-214 (2008-02-16)
A simple three-component chiral derivatization protocol for determining the enantiopurity of chiral primary amines by 1H NMR spectroscopic analysis is described here. The method involves condensation of the amines with 2-formylphenylboronic acid and enantiopure 1,1'-bi-2-naphthol. This approach affords a mixture
Shanshan Yu et al.
The Journal of organic chemistry, 76(8), 2814-2819 (2011-03-17)
The fluorescent properties of a series of H(8)BINOL-amine compounds are investigated. It is revealed that the intramolecular hydrogen bonds of these compounds contribute to the shift of the emission of their H(8)BINOL unit to a much longer wavelength. That is
Liheng Feng et al.
Organic & biomolecular chemistry, 9(8), 2938-2942 (2011-03-08)
A glucose sensing switch is formed by water soluble conjugated polymer (PP-S-BINOL) and boronic acid-functionalized benzyl viologen (o-BBV). The two-component system shows a high sensitivity for glucose sensing with a 17-fold increase in the fluorescence intensity in the presence of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service