Skip to Content
Merck
All Photos(2)

Key Documents

850489

Sigma-Aldrich

L-Tyrosine methyl ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH2CH(NH2)COOCH3 · HCl
CAS Number:
Molecular Weight:
231.68
Beilstein:
3917353
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

optical activity

[α]25/D +74.0°, c = 3 in pyridine

reaction suitability

reaction type: solution phase peptide synthesis

mp

192 °C (dec.) (lit.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

Cl.COC(=O)[C@@H](N)Cc1ccc(O)cc1

InChI

1S/C10H13NO3.ClH/c1-14-10(13)9(11)6-7-2-4-8(12)5-3-7;/h2-5,9,12H,6,11H2,1H3;1H/t9-;/m0./s1

InChI key

VXYFARNRGZWHTJ-FVGYRXGTSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesca Fontana et al.
Molecules (Basel, Switzerland), 26(2) (2021-01-14)
Configurationally stable 5-aza[6]helicene (1) was envisaged as a promising scaffold for non-conventional ionic liquids (IL)s. It was prepared, purified, and separated into enantiomers by preparative HPLC on a chiral stationary phase. Enantiomerically pure quaternary salts of 1 with appropriate counterions
Huseyin Bekir Yildiz et al.
Analytical chemistry, 80(8), 2811-2816 (2008-03-08)
The electrochemical and photoelectrochemical detection of tyrosinase (TR) activity (an indicative marker for melanoma cancer cells) is reported, using Pt nanoparticles (NPs) or CdS NPs as electrocatalytic labels or photoelectrochemical reporter units. The Pt NPs or CdS NPs are modified
M J Barjavel et al.
European journal of pharmacology, 258(3), 173-178 (1994-06-13)
The effects of the methyl esters of L-tyrosine (L-Tyr-OMe) and L-tryptophan (L-Trp-OMe) on the analgesic action of trans-3,4-dichloro-N-methyl-N-[2-(1-pyrrolidin)cyclohexyl]-benzene acetamide methane sulfonate (U-50,488H), a kappa-opioid receptor agonist, were determined in male Swiss-Webster mice using the tail-flick test. Intraperitoneal injections of U-50,488H
Separation of 125I-labelled prostaglandin E2-tyrosine methyl ester by reversed-phase high-performance liquid chromatography.
I Mucha et al.
Journal of chromatography, 438(1), 111-116 (1988-04-01)
M Tatsuta et al.
International journal of cancer, 48(5), 785-788 (1991-07-09)
The effect of tyrosine methyl ester (TME) on the incidence, number, and histological types of gastric cancers induced by N-methyl-N'-nitro-N-nitrosoguanidine (MNNG) was investigated in male Wistar rats. Rats were subcutaneously given TME, 512 mg/kg body weight, every other day after

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service