Skip to Content
Merck
All Photos(1)

Key Documents

806072

Sigma-Aldrich

bis-MPA-COOH dendrimer

trimethylol propane core, generation 4

Synonym(s):

G4-TMP-COOH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C423H566O282
Molecular Weight:
10162.85
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

solid

storage temp.

−20°C

SMILES string

O=C(OCC(COC(C(COC(C(COC(C(COC(C(COC(CCC(O)=O)=O)(COC(CCC(O)=O)=O)C)=O)(COC(C(COC(CCC(O)=O)=O)(COC(CCC(O)=O)=O)C)=O)C)=O)(COC(C(COC(C(COC(CCC(O)=O)=O)(COC(CCC(O)=O)=O)C)=O)(COC(C(COC(CCC(O)=O)=O)(COC(CCC(O)=O)=O)C)=O)C)=O)C)=O)(COC(C(COC(C(COC(C(COC(CCC(O)

InChI key

YDQBDELOMASXMM-UHFFFAOYSA-N

General description

  • End Group Functionality: Carboxyl surface group
  • Generation: 4
  • No. Surface Groups: 48
  • Calculated Mol. Wt.: 10162.9 g/mol
  • bis-MPA = 2,2-bis(hydroxymethyl)propionic acid

Application

Nontoxic dendrimers for drug delivery, with functional groups for conjugation or additional modifications.

Other Notes

InChI: 1S/C423H566O282/c1-47-423(234-703-375(610)420(44,228-697-369(604)414(38,216-685-357(592)402(26,192-661-333(568)378(2,144-613-285(520)96-48-237(424)425)145-614-286(521)97-49-238(426)427)193-662-334(569)379(3,146-615-287(522)98-50-239(428)429)147-616-288(523)99-51-240(430)431)217-686-358(593)403(27,194-663-335(570)380(4,148-617-289(524)100-52-241(432)433)149-618-290(525)101-53-242(434)435)195-664-336(571)381(5,150-619-291(526)102-54-243(436)437)151-620-292(527)103-55-244(438)439)229-698-370(605)415(39,218-687-359(594)404(28,196-665-337(572)382(6,152-621-293(528)104-56-245(440)441)153-622-294(529)105-57-246(442)443)197-666-338(573)383(7,154-623-295(530)106-58-247(444)445)155-624-296(531)107-59-248(446)447)219-688-360(595)405(29,198-667-339(574)384(8,156-625-297(532)108-60-249(448)449)157-626-298(533)109-61-250(450)451)199-668-340(575)385(9,158-627-299(534)110-62-251(452)453)159-628-300(535)111-63-252(454)455,235-704-376(611)421(45,230-699-371(606)416(40,220-689-361(596)406(30,200-669-341(576)386(10,160-629-301(536)112-64-253(456)457)161-630-302(537)113-65-254(458)459)201-670-342(577)387(11,162-631-303(538)114-66-255(460)461)163-632-304(539)115-67-256(462)463)221-690-362(597)407(31,202-671-343(578)388(12,164-633-305(540)116-68-257(464)465)165-634-306(541)117-69-258(466)467)203-672-344(579)389(13,166-635-307(542)118-70-259(468)469)167-636-308(543)119-71-260(470)471)231-700-372(607)417(41,222-691-363(598)408(32,204-673-345(580)390(14,168-637-309(544)120-72-261(472)473)169-638-310(545)121-73-262(474)475)205-674-346(581)391(15,170-639-311(546)122-74-263(476)477)171-640-312(547)123-75-264(478)479)223-692-364(599)409(33,206-675-347(582)392(16,172-641-313(548)124-76-265(480)481)173-642-314(549)125-77-266(482)483)207-676-348(583)393(17,174-643-315(550)126-78-267(484)485)175-644-316(551)127-79-268(486)487)236-705-377(612)422(46,232-701-373(608)418(42,224-693-365(600)410(34,208-677-349(584)394(18,176-645-317(552)128-80-269(488)489)177-646-318(553)129-81-270(490)491)209-678-350(585)395(19,178-647-319(554)130-82-271(492)493)179-648-320(555)131-83-272(494)495)225-694-366(601)411(35,210-679-351(586)396(20,180-649-321(556)132-84-273(496)497)181-650-322(557)133-85-274(498)499)211-680-352(587)397(21,182-651-323(558)134-86-275(500)501)183-652-324(559)135-87-276(502)503)233-702-374(609)419(43,226-695-367(602)412(36,212-681-353(588)398(22,184-653-325(560)136-88-277(504)505)185-654-326(561)137-89-278(506)507)213-682-354(589)399(23,186-655-327(562)138-90-279(508)509)187-656-328(563)139-91-280(510)511)227-696-368(603)413(37,214-683-355(590)400(24,188-657-329(564)140-92-281(512)513)189-658-330(565)141-93-282(514)515)215-684-356(591)401(25,190-659-331(566)142-94-283(516)517)191-660-332(567)143-95-284(518)519/h47-236H2,1-46H3,(H,424,425)(H,426,427)(H,428,429)(H,430,431)(H,432,433)(H,434,435)(H,436,437)(H,438,439)(H,440,441)(H,442,443)(H,444,445)(H,446,447)(H,448,449)(H,450,451)(H,452,453)(H,454,455)(H,456,457)(H,458,459)(H,460,461)(H,462,463)(H,464,465)(H,466,467)(H,468,469)(H,470,471)(H,472,473)(H,474,475)(H,476,477)(H,478,479)(H,480,481)(H,482,483)(H,484,485)(H,486,487)(H,488,489)(H,490,491)(H,492,493)(H,494,495)(H,496,497)(H,498,499)(H,500,501)(H,502,503)(H,504,505)(H,506,507)(H,508,509)(H,510,511)(H,512,513)(H,514,515)(H,516,517)(H,518,519)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Anna Carlmark et al.
Chemical Society reviews, 42(13), 5858-5879 (2013-05-01)
Dendritic polymers are highly branched, globular architectures with multiple representations of functional groups. These nanoscale organic frameworks continue to fascinate researchers worldwide and are today under intensive investigation in application-driven research. A large number of potential application areas have been

Related Content

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service