Skip to Content
Merck
All Photos(1)

Key Documents

806064

Sigma-Aldrich

bis-MPA-COOH dendrimer

trimethylol propane core, generation 2

Synonym(s):

G2-TMP-COOH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C99H134O66
CAS Number:
Molecular Weight:
2380.08
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

solid

storage temp.

−20°C

SMILES string

O=C(C(COC(C(COC(CCC(O)=O)=O)(COC(CCC(O)=O)=O)C)=O)(COC(C(COC(CCC(O)=O)=O)(COC(CCC(O)=O)=O)C)=O)C)OCC(CC)(COC(C(COC(C(COC(CCC(O)=O)=O)(COC(CCC(O)=O)=O)C)=O)(COC(C(COC(CCC(O)=O)=O)(COC(CCC(O)=O)=O)C)=O)C)=O)COC(C(COC(C(COC(CCC(O)=O)=O)(COC(CCC(O)=O)=O)C)=O)

InChI

1S/C99H134O66/c1-11-99(54-163-87(142)96(8,48-157-81(136)90(2,36-145-69(124)24-12-57(100)101)37-146-70(125)25-13-58(102)103)49-158-82(137)91(3,38-147-71(126)26-14-59(104)105)39-148-72(127)27-15-60(106)107,55-164-88(143)97(9,50-159-83(138)92(4,40-149-73(128)28-16-61(108)109)41-150-74(129)29-17-62(110)111)51-160-84(139)93(5,42-151-75(130)30-18-63(112)113)43-152-76(131)31-19-64(114)115)56-165-89(144)98(10,52-161-85(140)94(6,44-153-77(132)32-20-65(116)117)45-154-78(133)33-21-66(118)119)53-162-86(141)95(7,46-155-79(134)34-22-67(120)121)47-156-80(135)35-23-68(122)123/h11-56H2,1-10H3,(H,100,101)(H,102,103)(H,104,105)(H,106,107)(H,108,109)(H,110,111)(H,112,113)(H,114,115)(H,116,117)(H,118,119)(H,120,121)(H,122,123)

InChI key

SDMLAECJCDKFHK-UHFFFAOYSA-N

General description

  • End Group Functionality: Carboxyl surface group
  • Generation: 2
  • No. Surface Groups: 12
  • Calculated Mol. Wt.: 2380.1 g/mol
  • bis-MPA = 2,2-bis(hydroxymethyl)propionic acid

Application

Nontoxic dendrimers for drug delivery, with functional groups for conjugation or additional modifications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Self-assembled arrays of dendrimer-gold-nanoparticle hybrids for functional cell studies.
Anders Lundgren et al.
Angewandte Chemie (International ed. in English), 50(15), 3450-3453 (2011-03-12)
Chemical Society Reviews, 13, 4593-4613 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service