804630
Sodium-D-3-hydroxybutyrate-13C4
endotoxin tested, 99 atom % 13C, 99% (CP)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
13CH313CH(OH)13CH213CO2Na
Molecular Weight:
130.06
UNSPSC Code:
12352116
PubChem Substance ID:
Recommended Products
isotopic purity
99 atom % 13C
Assay
99% (CP)
form
powder
suitability
endotoxin tested
mass shift
M+4
storage temp.
2-8°C
SMILES string
[13CH3][13C@@H](O)[13CH2][13C](O[Na])=O
InChI
1S/C4H8O3.Na/c1-3(5)2-4(6)7;/h3,5H,2H2,1H3,(H,6,7);/q;+1/p-1/t3-;/m1./s1/i1+1,2+1,3+1,4+1;
InChI key
NBPUSGBJDWCHKC-UDJWXCBTSA-M
Related Categories
Application
For R&D use only. Not for drug,household, or other uses. Any other use is the sole responsibility of the enduser.Endotoxin testing is done on the bulk form of the product before subdivisionpackaging. This bulk test does not guarantee that the subdivision or repackagedaliquot is endotoxin-free when it is received or used by the customer and doesnot imply suitability for any particular purpose. If the product must beendotoxin-free for the intended application, the product should be tested priorto actual use.
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service