Skip to Content
Merck
All Photos(1)

Key Documents

802913

Sigma-Aldrich

(24R),24,25-Dihydroxyvitamin D3-26,26,26,27,27,27-d6 solution

100 μg/mL in ethanol, 98 atom % D, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27D6H38O3
Molecular Weight:
422.67
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

97% (CP)

form

solution

concentration

100 μg/mL in ethanol

technique(s)

mass spectrometry (MS): suitable

color

colorless

application(s)

detection

mass shift

M+6

storage temp.

−20°C

SMILES string

C[C@H](CC[C@H](O)C(C([2H])([2H])[2H])(C([2H])([2H])[2H])O)C1CCC([C@]1(C)CCC/2)C2=C\C=C(C[C@@H](O)CC3)/C3=C

InChI

1S/C27H44O3/c1-18-8-12-22(28)17-21(18)11-10-20-7-6-16-27(5)23(13-14-24(20)27)19(2)9-15-25(29)26(3,4)30/h10-11,19,22-25,28-30H,1,6-9,12-17H2,2-5H3/b20-10+,21-11-/t19-,22+,23?,24?,25+,27-/m1/s1/i3D3,4D3

InChI key

FCKJYANJHNLEEP-PRFFRHJRSA-N

Related Categories

General description

(24R)-24,25-Dihydroxyvitamin D3 is a metabolite of vitamin D3. It is known to increase bone volume and mechanical strength.
(24R),24,25-Dihydroxyvitamin D3-26,26,26,27,27,27-d6 is a deuterated metabolite of vitamin D3 wherein methyl protons of C-26, C-27 carbons are replaced by deuterium.

Application

(24R),24,25-Dihydroxyvitamin D3-26,26,26,27,27,27-d6 can be used as an analytical standard for accurate data interpretation in specific biological samples.

Packaging

1 mL in ampule
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F

Flash Point(C)

14 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of C-3 epimerization in (24R)-24, 25-dihydroxyvitamin D3 catalyzed by hydroxysteroid dehydrogenase
Higashi T, et al.
Journal of Pharmaceutical and Biomedical Analysis, 36(3), 429-436 (2004)
24R, 25-dihydroxyvitamin D3: an essential vitamin D3 metabolite for both normal bone integrity and healing of tibial fracture in chicks
Seo E, et al.
Endocrinology, 138(9), 3864-3872 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service