705470
Vitamin K-d7 (5,6,7,8-d4, 2-methyl-d3)
99 atom % D, 97% (CP Sum of E & Z Isomers)
Synonym(s):
2-methyl-3-phytyl-1,4-naphthoquinone--d7 (5,6,7,8-d4, 2-methyl-d3), 3-Phytylmenadione-d7 (5,6,7,8-d4, 2-methyl-d3), Phylloquinone-d7 (5,6,7,8-d4, 2-methyl-d3), Vitamin K1-d7 (5,6,7,8-d4, 2-methyl-d3)
About This Item
Recommended Products
isotopic purity
99 atom % D
Quality Level
Assay
97% (CP Sum of E & Z Isomers)
form
liquid
technique(s)
mass spectrometry (MS): suitable
color
yellow to orange
mp
-20 °C
density
0.999 g/mL at 25 °C
mass shift
M+7
storage temp.
−20°C
SMILES string
[2H]c1c([2H])c([2H])c2C(=O)C(=C(C\C=C(/C)CCC[C@H](C)CCC[C@H](C)CCCC(C)C)C(=O)c2c1[2H])C([2H])([2H])[2H]
InChI
1S/C31H46O2/c1-22(2)12-9-13-23(3)14-10-15-24(4)16-11-17-25(5)20-21-27-26(6)30(32)28-18-7-8-19-29(28)31(27)33/h7-8,18-20,22-24H,9-17,21H2,1-6H3/b25-20+/t23-,24-/m1/s1/i6D3,7D,8D,18D,19D
InChI key
MBWXNTAXLNYFJB-NYKMUXSXSA-N
General description
Application
Packaging
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service