Skip to Content
Merck
All Photos(1)

Key Documents

797685

Sigma-Aldrich

ADIBO-PEG4-acid

90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H36N2O8
CAS Number:
Molecular Weight:
552.62
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

90% (HPLC)

form

liquid

reaction suitability

reaction type: click chemistry
reagent type: linker

functional group

carboxylic acid

storage temp.

−20°C

SMILES string

O=C(CCC(NCCOCCOCCOCCOCCC(O)=O)=O)N1CC2=C(C=CC=C2)C#CC3=C1C=CC=C3

InChI

1S/C30H36N2O8/c33-28(31-14-16-38-18-20-40-22-21-39-19-17-37-15-13-30(35)36)11-12-29(34)32-23-26-7-2-1-5-24(26)9-10-25-6-3-4-8-27(25)32/h1-8H,11-23H2,(H,31,33)(H,35,36)

InChI key

LKSAMQQFMTVPKD-UHFFFAOYSA-N

Application

Acid functionalized cyclooctyne derivative. Cyclooctynes are useful in strain-promoted copper-free azide-alkyne cycloaddition reactions. This dibenzocyclooctyne will react with azide functionalized compounds or biomolecules without the need for a Cu(I) catalyst to result in a stable triazole linkage. The PEG spacer adds hydrophilicity to the product to decrease aggregation.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service