Skip to Content
Merck
All Photos(1)

Key Documents

791539

Sigma-Aldrich

Titania paste, reflector

Synonym(s):

Greatcell Solar® WER2-O, TiO2 paste

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

description

Crystal Structure: > 99% anatase (analysis carried out on starting material, prior to paste manufacture)

Quality Level

form

paste (white)

concentration

20.0 wt. % (+/- 1.2 wt%)

avg. part. size

150-250 nm (scatter)

viscosity

15000-25000 mPa.s (Analysis carried out with 20mm 4 degree cone/plate; 40 s-1)

SMILES string

[Ti](=O)=O

InChI

1S/2O.Ti

InChI key

GWEVSGVZZGPLCZ-UHFFFAOYSA-N

General description

Titania paste, the reflector is a precursor for titania, that forms a mesoporous light scattering layer on the fluorine doped tin oxide (FTO) substrates. It is used in a variety of electrochemical devices to improve electron mobility and power efficiency.[1][2]

Application

Light scattering titania paste can be coated on the primary transparent layer of TiO2 to form a bilayered titanium film. It can be used as a negative electrode for dye sensitized solar cells (DSSCs).[3][4][5]
Scattering Reflector Titania Paste can be used in conjunction with Transparent Titania Paste for enhanced DSSC performance in applications where transparency is not required.
Scattering Titania Paste contains highly dispersed anatase scattering particles (150nm to 250nm). This paste produces a non-active scattering layer when applied onto a pre-printed active layer.
After drying; this paste must be fired – together with the pre-printed active layers – at or above 500°C. This results in an opaque sintered layer; with a thickness of ~3μm for one printed layer; when using a 90T mesh screen.

Storage: Store in the dark at 20°C

Legal Information

Product of Greatcell Solar® exclusive supplier of Sun2 products under license from EPFL.
Greatcell Solar® is a registered trademark of Greatcell Solar
Greatcell Solar is a registered trademark of Greatcell Solar

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

195.8 °F

Flash Point(C)

91 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pt-free spray coated reduced graphene oxide counter electrodes for dye sensitized solar cells
Nagavolu C, et al.
Solar Energy, 137(11), 143-147 (2016)
CdSe-sensitized mesoscopic TiO 2 solar cells exhibiting> 5% efficiency: redundancy of CdS buffer layer
Hossain MA, et al.
Journal of Materials Chemistry, 22(32), 16235-16242 (2012)
Joint electrical, photophysical and computational studies on D-pi-A dye sensitized solar cells: the impacts of dithiophene rigidification
Xu M, et al.
Chemical Science, 3(4), 976-983 (2012)
Band engineered ternary solid solution CdS x Se 1- x-sensitized mesoscopic TiO 2 solar cells
Hossain MA, et al.
Physical Chemistry Chemical Physics, 14(19), 7154-7161 (2012)
Electrical and photophysical analyses on the impacts of arylamine electron donors in cyclopentadithiophene dye-sensitized solar cells
Xu M, et al.
Energy & Environmental Science, 4(11), 4735-4742 (2011)

Articles

Few Monolayer Atomic Layer Deposition (ALD) on Surfaces and Interfaces for Energy Applications

Dye-sensitized solar cells (DSCs) are 3rd generation solar cells combining the promise of high efficiency with low production costs.

Organic photovoltaics (OPVs) represent a low-cost, lightweight, and scalable alternative to conventional solar cells. While significant progress has been made in the development of conventional bulk heterojunction cells, new approaches are required to achieve the performance and stability necessary to enable commercially successful OPVs.

While dye sensitization as the basis for color photography has been accepted for a very long time,1 attempts to use this principle for the conversion of solar light to electricity generally had resulted only in very low photocurrents, below 100 nA/cm

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service