Skip to Content
Merck
All Photos(1)

Key Documents

779105

Sigma-Aldrich

β-Cyclodextrin

produced by Wacker Chemie AG, Burghausen, Germany, ≥95.0% (HPLC)

Synonym(s):

Cavamax® W7, beta-Cyclodextrin, Caraway, Cycloheptaamylose, Cyclomaltoheptaose, Schardinger β-Dextrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H70O35
CAS Number:
Molecular Weight:
1134.98
Beilstein:
78623
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

grade

produced by Wacker Chemie AG, Burghausen, Germany

Quality Level

Assay

≥95.0% (HPLC)

form

powder

optical activity

[α]20/D +162±3°, c = 1.5% in H2O

loss

≤14.0% loss on drying

mp

290-300 °C (dec.) (lit.)

functional group

ether
hydroxyl

SMILES string

OC[C@H]1O[C@@H]2O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)O[C@H]6[C@H](O)[C@@H](O)[C@H](O[C@@H]6CO)O[C@H]7[C@H](O)[C@@H](O)[C@H](O[C@@H]7CO)O[C@H]8[C@H](O)[C@@H](O)[C@H](O[C@@H]8CO)O[C@H]1[C@H](O)[C@H]2O

InChI

1S/C42H70O35/c43-1-8-29-15(50)22(57)36(64-8)72-30-9(2-44)66-38(24(59)17(30)52)74-32-11(4-46)68-40(26(61)19(32)54)76-34-13(6-48)70-42(28(63)21(34)56)77-35-14(7-49)69-41(27(62)20(35)55)75-33-12(5-47)67-39(25(60)18(33)53)73-31-10(3-45)65-37(71-29)23(58)16(31)51/h8-63H,1-7H2/t8-,9-,10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-/m1/s1

InChI key

WHGYBXFWUBPSRW-FOUAGVGXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

β-Cyclodextrin, a water-soluble cyclic oligosaccharide consisting of seven α-(1,4)-linked glucopyranose subunits, is used in the preparation of:
  • Linear cationic β-cyclodextrin-based polymers (β-CDPs) and polycationic β-cyclodextrin ‘click clusters′ for gene delivery applications.
  • Multiresponsive supramolecular nanogated ensemble by grafting onto mesoporous silica for controlled release of trapped molecules.
  • EDTA-cross-linked β-cyclodextrin (EDTA-β-CD) bifunctional adsorbent for simultaneous adsorption of metals and dyes.
  • β-Cyclodextrin derived siRNA delivery vector for gene silencing.
  • β-CD chiral stationary phase (CCNCSP) for the enantioseparation of chiral drugs by high-performance liquid chromatography (HPLC).

Legal Information

Cavasol is a registered trademark of Wacker Chemie AG

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclodextrins
Encyclopedia of Polymer Science and Technology, Concise (2004)
Polycationic β-cyclodextrin ?click clusters?: monodisperse and versatile scaffolds for nucleic acid delivery
Srinivasachari S, et al.
Journal of the American Chemical Society, 130(14), 4618-4627 (2008)
Multiresponsive supramolecular nanogated ensembles
Liu R, et al.
Journal of the American Chemical Society, 131(42), 15128-15129 (2009)
EDTA-cross-linked β-cyclodextrin: an environmentally friendly bifunctional adsorbent for simultaneous adsorption of metals and cationic dyes
Zhao F and Repo E
Environmental Science & Technology, 49(17), 10570-10580 (2015)
Effects of structure of β-cyclodextrin-containing polymers on gene delivery
Hwang SJ, et al.
Bioconjugate Chemistry, 12(2), 280-290 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service