Skip to Content
Merck
All Photos(1)

Key Documents

771953

Sigma-Aldrich

exo-5-Norbornene-2-methanol

≥99% (exo)

Synonym(s):

exo-5-Norbornenyl methyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12O
CAS Number:
Molecular Weight:
124.18
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99% (exo)

form

liquid

refractive index

n20/D 1.499

bp

200-205/760 mmHg

density

1.030 at 25 °C

SMILES string

OCC1C[C@@H]2C[C@H]1C=C2

InChI

1S/C8H12O/c9-5-8-4-6-1-2-7(8)3-6/h1-2,6-9H,3-5H2/t6-,7+,8?/m0/s1

InChI key

LUMNWCHHXDUKFI-KJFJCRTCSA-N

General description

Exo-5-Norbornene-2-methanol is a norbornene derivative which is widely used as a monomer in the synthesis of polynorbornenes by vinyl polymerisation or ring opening methathesis polymerisation (ROMP). They are also used as a monomer or an intermediate in the synthesis of photoresist materials. This material is also widely used as an intermediate in the synthesis of pharmaceutically and biologically active compounds.

Application

Used as a monomer in the synthesis of polynorbornenes by ring opening metathesis polymerisation.

Features and Benefits

  • Produces polymers with high thermal stability and transparency which are useful for optical applications.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

195.0 °F

Flash Point(C)

90.55 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Stereo-Selective Synthesis of 5-Norbornene-2-exo-carboxylic Acid-Rapid Isomerization and Kinetically Selective Hydrolysis.
Kanao, M., Otake, A., Tsuchiya, K., & Ogino, K.
International Journal of Organic Chemistry, 2, 26-30 (2012)
Water-soluble polymers from acid-functionalized norbornenes
Lienkamp, K., Kins, C. F., Alfred, S. F., Madkour, A. E., & Tew, G. N.
Journal of Polymer Science Part A: Polymer Chemistry, 47(5), 1266-1273 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service