Skip to Content
Merck
All Photos(1)

Key Documents

248533

Sigma-Aldrich

5-Norbornene-2-methanol

98%, mixture of endo and exo

Synonym(s):

2-(Hydroxymethyl)bicyclo[2.2.1]hept-5-ene, 2-Hydroxymethyl-5-norbornene, 2-Norbornen-5-methanol, 5-(Hydroxymethyl)bicyclo[2.2.1]hept-2-ene, 5-Hydroxymethyl-2-norbornene, Bicyclo[2.2.1]hept-5-en-2-ylmethanol, Norbornene-5-methyl alcohol, [Bicyclo[2.2.1]hept-5-en-2-yl]methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12O
CAS Number:
Molecular Weight:
124.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.500 (lit.)

bp

97 °C/20 mmHg (lit.)

density

1.027 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

OCC1C[C@@H]2C[C@H]1C=C2

InChI

1S/C8H12O/c9-5-8-4-6-1-2-7(8)3-6/h1-2,6-9H,3-5H2/t6-,7+,8?/m0/s1

InChI key

LUMNWCHHXDUKFI-KJFJCRTCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Co-polymerization of 5-norbornene-2-methanol with ethylene using heterogeneous silica supported catalysts has been reported.

Application

5-Norbornene-2-methanol has been used in the preparation of:
  • 4-(norborn-2-ene-5-ylmethyleneoxy)-2,2′:6′,2′′-terpyridine
  • 5- {[(3,5-di-t-butyl-4-hydroxy-benzoyl)oxy]methyl}-2-norbornene (BHBN)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhen Zhang et al.
Polymers, 12(4) (2020-04-16)
Injectable, covalently cross-linked hydrogels have been widely investigated in drug delivery systems due to their superior mechanical properties and long-term stability. Conventional covalently cross-linked hydrogels are formed by chemical reactions that may interfere with natural biochemical processes. In this work
Homogeneous and heterogeneous metallocene/MAO-catalyzed polymerization of functionalized olefins.
Goretzki R and Fink G.
Macromolecular Chemistry and Physics, 200(4), 881-886 (1999)
Synthesis of a polynorbornene containing 3, 5-di-TERT-butyl-4-hydroxybenzoyl group.
Tienkopatchev MA, et al.
Polymer Bull., 34(4), 385-391 (1995)
Access to heterogeneous atom-transfer radical polymerization (ATRP) catalysts based on dipyridylamine and terpyridine via ring-opening metathesis polymerization (ROMP).
Kroll R, et al.
Macromolecular Chemistry and Physics, 202(5), 645-653 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service