Skip to Content
Merck
All Photos(1)

Key Documents

753092

Sigma-Aldrich

Pentafluorophenyl acrylate

contains <200 ppm monomethyl ether hydroquinone as inhibitor, 98%

Synonym(s):

2-Propenoic acid pentafluorophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H3F5O2
CAS Number:
Molecular Weight:
238.11
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

liquid

contains

<200 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n/D 1.434

density

1.446

storage temp.

2-8°C

SMILES string

Fc1c(F)c(F)c(OC(=O)C=C)c(F)c1F

InChI

1S/C9H3F5O2/c1-2-3(15)16-9-7(13)5(11)4(10)6(12)8(9)14/h2H,1H2

InChI key

RFOWDPMCXHVGET-UHFFFAOYSA-N

Application

Acrylate monomer with an activated ester for coupling (esterification/amidation) reactions
Pentafluorophenyl acrylate may be used in the preparation of highly porous polymers (poly HIPs). Pentafluorophenyl acrylate (PFPA) can undergo reversible addition fragmentation transfer (RAFT) polymerization with oligoethylene glycol acrylate (OEGA) or diethylene glycol acrylate (DEGA).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Factors influencing the synthesis and the post-modification of PEGylated pentafluorophenyl acrylate containing copolymers
Beija M, et al.
European Polymer Journal, 49, 3060?3071-3060?3071 (2013)
Reactive thiol-ene emulsion-templated porous polymers incorporating pentafluorophenyl acrylate
Kircher L, et al.
Polymer, 54(7), 1755-1761 (2013)
Yuwaporn Pinyakit et al.
Journal of materials chemistry. B, 8(3), 454-464 (2019-12-14)
Recently, pH-responsive polymeric micelles have gained significant attention as effective carriers for anti-cancer drug delivery. Herein, pH-responsive polymeric micelles were constructed by a simple post-polymerization modification of a single homopolymer, poly(pentafluorophenyl acrylate) (PPFPA). The PPFPA was first subjected to modification

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service