Skip to Content
Merck
All Photos(1)

Key Documents

741108

Sigma-Aldrich

Pentafluorophenyl methacrylate

contains MEHQ as inhibitor, 95%

Synonym(s):

Methacrylic acid, pentafluorophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H5F5O2
CAS Number:
Molecular Weight:
252.14
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

95%

form

liquid

contains

MEHQ as inhibitor

refractive index

n20/D 1.438

density

1.394 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)Oc1c(F)c(F)c(F)c(F)c1F

InChI

1S/C10H5F5O2/c1-3(2)10(16)17-9-7(14)5(12)4(11)6(13)8(9)15/h1H2,2H3

InChI key

NIJWSVFNELSKMF-UHFFFAOYSA-N

Application

The PFP unit acts as an activated ester for coupling (esterification/amidation) reactions. This is also a monomer for low refractive index polymers (n ~1.40).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

172.0 °F

Flash Point(C)

77.8 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Copolymerization of methyl methacrylate with pentafluorophenyl methacrylate and physical characteristics of the polymers (author's transl)].
Y Kadoma et al.
Iyo Kizai Kenkyujo hokoku. Reports of the Institute for Medical and Dental Engineering, Tokyo Medical and Dental University, 15, 23-29 (1981-01-01)
L Francesch et al.
Langmuir : the ACS journal of surfaces and colloids, 23(7), 3927-3931 (2007-03-07)
Pulsed-plasma polymerization has been used to deposit ultrathin layers of pentafluorophenyl methacrylate by using low duty cycles and low power input. The monomer structure can be retained such that the chemical reactivity of the active ester group could be studied
Claudia Battistella et al.
Biomacromolecules, 18(6), 1855-1865 (2017-04-15)
Inhibition of P-glycoprotein (P-gp) transporter is an attractive approach for the reversion of cancer-associated multidrug resistance (MDR). Poly(N-(2-hydroxypropyl) methacrylamide) (PHPMA)-based carriers that are able to release the anticancer drug doxorubicin in the lysosomes have shown promise to reduce P-gp mediated
Nikhil K Singha et al.
Biomacromolecules, 12(8), 2908-2913 (2011-07-08)
Herein the concept of tandem postpolymerization modification as a versatile route to synthesize well-defined, highly functionalized polymers is introduced. Poly(pentafluorophenyl methacrylate) obtained by atom transfer radical polymerization was first modified with allylamine, which displaces the active ester to give well-defined
Luis Duque et al.
Biomacromolecules, 11(10), 2818-2823 (2010-09-14)
Thin films of plasma polymerized pentafluorophenyl methacrylate (pp-PFM) offer highly reactive ester groups throughout the structure of the film that allow for subsequent reactions with different aminated reagents and biological molecules. The present paper follows on from previous work on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service