752169
tert-Butyl diazoacetate solution
15% in toluene
Synonym(s):
2-Diazoacetic acid tert-butyl ester solution
About This Item
Recommended Products
form
liquid
Quality Level
concentration
15% in toluene
refractive index
n20/D 1.490
density
0.875 g/mL at 25 °C
storage temp.
2-8°C
SMILES string
CC(C)(C)OC(=O)C=[N+]=[N-]
InChI
1S/C6H10N2O2/c1-6(2,3)10-5(9)4-8-7/h4H,1-3H3
InChI key
JBVSBLLOZVDAAZ-UHFFFAOYSA-N
Related Categories
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3
Target Organs
Central nervous system, Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
37.0 °F - closed cup
Flash Point(C)
2.8 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service