Skip to Content
Merck
All Photos(3)

Key Documents

749141

Sigma-Aldrich

Iron(III) oxo acetate perchlorate hydrate

greener alternative

Synonym(s):

Hexakis[μ-(acetato-κO:κO′ )]triaqua-μ3-oxotriiron(1+) perchlorate hydrate, Trimeric Fe cluster

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H24Fe3O16 · ClO4 · xH2O
Molecular Weight:
691.29 (anhydrous basis)
UNSPSC Code:
26111700
NACRES:
NA.23

form

powder

Quality Level

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Pictograms

Flame over circle

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steffen Hausdorf et al.
Dalton transactions (Cambridge, England : 2003), 7(7), 1107-1113 (2009-03-27)
An ethynylene diisophthalic acid linker molecule was synthesized and used to form a zinc carboxylate-based metal organic framework (MOF) with very large pores and unit cell volume resulting from the unusual combination of structurally different inorganic units forming the secondary
Steffen Hausdorf et al.
The journal of physical chemistry. A, 112(33), 7567-7576 (2008-07-26)
The contributions of terephthalic acid and Zn(2+)-coordinated water in N,N-diethylformamide (DEF) to the overall proton activity in the synthesis of MOF-5 (Zn4O(BDC)3, BDC = 1,4-benzenedicarboxylate) were quantitatively determined by combined electrochemical and UV-vis spectroscopic measurements. Structural transformations of zinc carboxylate-based
Steffen Hausdorf et al.
Journal of the American Chemical Society, 132(32), 10978-10981 (2010-08-12)
A simple two-component procedure was developed to synthesize not only classical zinc-based IRMOFs represented by MOF-5 but also the cobalt and beryllium homologues of this most prominent MOF. The procedure is the first manifestation of mirroring the IRMOF series with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service