747858
4-Fluoro-1H-pyrazole
95%
Synonym(s):
4-Fluoropyrazole
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C3H3FN2
CAS Number:
Molecular Weight:
86.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
form
solid
mp
38-40 °C
functional group
fluoro
SMILES string
FC1=CNN=C1
InChI
1S/C3H3FN2/c4-3-1-5-6-2-3/h1-2H,(H,5,6)
InChI key
RYXJXPHWRBWEEB-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
One-pot synthesis of. beta.-keto sulfones and. beta.-keto sulfoxides from carboxylic acids.
Ibarra CA, et al.
The Journal of Organic Chemistry, 54(23), 5620-5623 (1989)
Fluoromalonaldehyde bis (dialkyl acetals): synthesis by carbene condensation and transformation to dialkyl fluoromalonates and fluorinated heterocyclic compounds.
Molines H and Wakselman C.
The Journal of Organic Chemistry, 54(23), 5618-5620 (1989)
Relationship between substituent-induced energy and charge effects in proton-transfer equilibria involving heteroaromatic nitrogen systems. The lone pair charge approach.
Catalan J, et al.
Journal of the American Chemical Society, 106(22), 6552-6556 (1984)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service