Skip to Content
Merck
All Photos(1)

Key Documents

71208

Sigma-Aldrich

Sodium ethanethiolate

technical, ~90% (RT)

Synonym(s):

Ethanethiol sodium salt, Ethyl mercaptan sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2SNa
CAS Number:
Molecular Weight:
84.12
Beilstein:
3593647
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

~90% (RT)

form

crystals

SMILES string

CCS[Na]

InChI

1S/C2H6S.Na/c1-2-3;/h3H,2H2,1H3;/q;+1/p-1

InChI key

QJDUDPQVDAASMV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium ethanethiolate is a reagent generally used to synthesize various thioethers. It can be employed as a ligand in the preparation of organometallic complexes. It is also used in the synthesis of thiolated amino-alcohols.

Other Notes

Alkanethiolates are efficient reagents for SN2 dealkylation of esters and aryl ethers

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G.I. Feutrill et al.
Tetrahedron Letters, 1327-1327 (1970)
P.A. Bartlett et al.
Tetrahedron Letters, 4459-4459 (1970)
Novel thiolated amino-alcohols as chiral ligands for copper-catalyzed asymmetric nitro-aldol reactions.
Mansawat, Woraluk et al.
Tetrahedron Letters, 48(24), 4235-4238 (2007)
Kojyl thioether derivatives having both tyrosinase inhibitory and anti-inflammatory properties.
Rho, Ho Sik et al.
Bioorganic & Medicinal Chemistry Letters, 20(22), 6569-6571 (2010)
T.R. Kelly et al.
Tetrahedron Letters, 3859-3859 (1977)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service