Skip to Content
Merck
All Photos(1)

Key Documents

743844

Sigma-Aldrich

Paraldehyde

Arxada quality, ≥99.5% (GC)

Synonym(s):

2,4,6-Trimethyl-1,3,5-trioxacyclohexane, s-Trimethyltrioxymethylene, Acetaldehyde trimer, NSC 9799, Paracetaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O3
CAS Number:
Molecular Weight:
132.16
Beilstein:
80142
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.5% (GC)

form

liquid

quality

Arxada quality

manufacturer/tradename

Arxada AG

drug control

USDEA Schedule IV

impurities

≤0.10% water
≤0.30% acetic acid

density

0.994 g/mL at 20 °C (lit.)

SMILES string

CC1OC(C)OC(C)O1

InChI

1S/C6H12O3/c1-4-7-5(2)9-6(3)8-4/h4-6H,1-3H3

InChI key

SQYNKIJPMDEDEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Modified Sanford reaction
  • Synthesis of N-arylbenzoquinaldinium derivatives with antimicrobial activity
  • Hydroformylation of alkenylamines
  • Studies of molecules with inhibitory activity against HIV-1 integrase
  • Irregular Wittig reactions for the formation of α-hydroxyketones

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmacological management of convulsive status epilepticus in children.
R C Scott et al.
Developmental medicine and child neurology, 41(3), 207-210 (1999-04-21)
Will Townend et al.
Emergency medicine journal : EMJ, 19(1), 50-50 (2002-01-05)
A short cut review was carried out to establish whether phenytoin or paraldehyde should be given as the second drug for resistant fits in children. Altogether 41 papers were found using the reported search, of which none answered the question
Audit of treatment of convulsive status epilepticus.
Jithangi Wanigasinghe et al.
The Ceylon medical journal, 53(3), 110-110 (2008-11-06)
Serum creatine kinase levels after a single intramuscular injection--dependence on injection volume.
Z Azzam et al.
Annals of clinical biochemistry, 31 ( Pt 2), 193-194 (1994-03-01)
J B Messick et al.
Infection and immunity, 61(9), 3803-3810 (1993-09-01)
The binding, internalization, and proliferation of Ehrlichia risticii in P388D1 cells and equine polymorphonuclear (PMN) leukocytes were studied by immunofluorescent staining and flow cytometric analysis. The binding of ehrlichiae to P388D1 cells at 4 degrees C was dose dependent, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service