Skip to Content
Merck
All Photos(1)

Key Documents

743038

Sigma-Aldrich

Sodium anthraquinone-2-sulfonate

≥98% (HPLC)

Synonym(s):

9,10-Dihydro-9,10-dioxo-2-anthracenesulfonic acid sodium salt, Anthraquinone-2-sulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H7NaO5S
CAS Number:
Molecular Weight:
310.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98% (HPLC)

form

powder

SMILES string

[Na+].[O-]S(=O)(=O)c1ccc2C(=O)c3ccccc3C(=O)c2c1

InChI

1S/C14H8O5S.Na/c15-13-9-3-1-2-4-10(9)14(16)12-7-8(20(17,18)19)5-6-11(12)13;/h1-7H,(H,17,18,19);/q;+1/p-1

InChI key

GGCZERPQGJTIQP-UHFFFAOYSA-M

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaofeng Xie et al.
Journal of chromatography. A, 1523, 242-247 (2017-08-19)
The design of a miniaturized LED-based UV-absorption detector was significantly improved for on-column nanoflow LC. The detector measures approximately 27mm×24mm×10mm and weighs only 30g. Detection limits down to the nanomolar range and linearity across 3 orders of magnitude were obtained
Romina R Heymann et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 16(6), 1003-1008 (2017-05-13)
Photolysis of anthraquinone or flavin photosensitizers in the presence of calcium EDTA complexes results in decomposition of the EDTA complex, releasing free Ca
Bin Huang et al.
Ecotoxicology and environmental safety, 171, 313-320 (2019-01-07)
Photolysis and microbial activity are relatively obvious in shallow, eutrophic waters with low dissolved oxygen content. Ubiquitous humic acid (HA) can act as electron acceptor and be reduced by bacterial under such conditions, and the reduced form of humic acid
Thuy P Nguyen et al.
The Analyst, 145(14), 5041-5052 (2020-06-20)
Carbon film electrodes can often be used without pretreatment, and their fabrication allows for flexibility in size and shape and for mass production. In this work, we are exploring layered structures comprised of thin films of carbon on gold (eC/Au)
Xiaoling He et al.
Journal of applied biomaterials & functional materials, 14(4), e404-e412 (2016-11-04)
The aim of this study was to develop a novel thermoresponsive material suited for tissue engineering and investigate the growth and harmless detachment of cells cultured on the surface of thermoresponsive tissue culture polystyrene (TCPS). Thermoresponsive N-isopropylacrylamide (NIPAAm) and biocompatible

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service