74291
2-(Fmoc-amino)ethyl bromide
≥95.0% (AT)
Synonym(s):
9-Fluorenylmethyl N-(2-bromoethyl)carbamate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C17H16BrNO2
CAS Number:
Molecular Weight:
346.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥95.0% (AT)
form
powder
reaction suitability
reagent type: cross-linking reagent
functional group
Fmoc
bromo
storage temp.
2-8°C
SMILES string
BrCCNC(OCC1C(C=CC=C2)=C2C3=C1C=CC=C3)=O
InChI
1S/C17H16BrNO2/c18-9-10-19-17(20)21-11-16-14-7-3-1-5-12(14)13-6-2-4-8-15(13)16/h1-8,16H,9-11H2,(H,19,20)
InChI key
GDIPNIOJNLDDRP-UHFFFAOYSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service