Skip to Content
Merck
All Photos(1)

Key Documents

15369

Sigma-Aldrich

N-Boc-ethylenediamine

≥98.0% (NT)

Synonym(s):

N-Boc-1,2-diaminoethane, tert-Butyl N-(2-aminoethyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCONHCH2CH2NH2
CAS Number:
Molecular Weight:
160.21
Beilstein:
1932330
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (NT)

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.458 (lit.)
n20/D 1.458

bp

72-80 °C/0.1 mmHg (lit.)

density

1.012 g/mL at 20 °C (lit.)

functional group

Boc
amine

SMILES string

NCCNC(OC(C)(C)C)=O

InChI

1S/C7H16N2O2/c1-7(2,3)11-6(10)9-5-4-8/h4-5,8H2,1-3H3,(H,9,10)

InChI key

AOCSUUGBCMTKJH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Facile synthesis of new N-(aminocycloalkylene) amino acid compounds: Describes the use of N-Boc-ethylenediamine in the preparation of benzyl 2-{[2-(Boc-amino)ethyl]amino}acetate (G Matulevičiūtė et al., 2023).
  • Synthesis of a new tripod BODIPY dye: Reports on the chemical synthesis involving N-Boc-ethylenediamine for creating a BODIPY dye (H Wang et al., 2022).
  • Single‐Entity Electrocatalysis: Discusses the application of N-Boc-ethylenediamine in modifying Co3O4 nanoparticles used in electrocatalysis (T Quast et al., 2021).

Other Notes

Mono-protected derivative of ethylenediamine, preparation of pharmacologically active analogues; introduction of an ethylenediamine spacer.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of N,N-dilactitol ethylenediamine: a versatile spacer for attachment of residualizing labels to protein.
M M Dowd et al.
Analytical biochemistry, 205(2), 369-371 (1992-09-01)
W S Saari et al.
Journal of medicinal chemistry, 34(10), 3132-3138 (1991-10-01)
Basic nitrobenzenesulfonamides containing nitroisopropyl and (ureidooxy)methyl groups were prepared and evaluated as novel hypoxic cell selective cytotoxic agents. In vitro, N-(2-aminoethyl)-N-methyl-3-nitro-4-(1-methyl-1-nitroethyl)benzene sulfonamide hydrochloride (11) proved to be preferentially toxic to hypoxic EMT6 mammary carcinoma cells. At 1 mM concentration in
Hao Li et al.
Investigative ophthalmology & visual science, 58(12), 5142-5150 (2017-10-08)
Intravitreal injection of antiangiogenic agents is becoming a standard treatment for neovascular retinal diseases. Sustained release of therapeutics by injecting colloidal carriers is a promising approach to reduce the injection frequency, which reduces treatment burdens and the risk of complications
Qi Zhang et al.
Journal of biophotonics, 11(6), e201700339-e201700339 (2018-01-18)
Targeting cyclooxygenase-2 (COX-2) for molecular imaging is an attractive approach applicable for its overexpression in inflammation and many malignancies. Herein, for monitoring COX-2, we synthesize a specific COX-2 probe celecoxib-MPA probe (CMP), based on celecoxib and a water-soluble near-infrared dye
G. Kada et al.
Biochimica et Biophysica Acta, 1427, 33-33 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service